Would you like to log in again?
CFAVax, OZ Biosciences
Supplier: OZ Biosciences
CFA0010 CFA0100 CFA0050
103258-204EA 71.08 USD InStock
103258-204 103258-208 103258-206
CFAVax, OZ Biosciences
Media for Microbiology Additives and Supplements for Microbiology
Complete Freund’s Adjuvant (CFA) is a water-in-oil emulsion containing 1 mg per mL heat-killed Mycobacterium tuberculosis.
CFAVax consists of a mixture of mineral oil and emulsifier in a ratio of 85% v/v oil and 15% v/v emulsifier. Each 10mL vial contains 10 mg mycobacterium. Importantly, CFAVax is not a pre-formed emulsion and thus it must be mixed with an equal volume of aqueous solution of antigen and subsequently emulsified prior to use. The emulsifier is mannide monooleate, an ester consisting of mannitol as the hydrophilic residue and oleic acid (unsaturated C18 fatty acid) as the hydrophobic part.
Delivery information: Shipped at room temperature
Packaging: Store at +4°C
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty