Would you like to log in again?
3D-Fect™ Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
3D-Fect™
TF21000 TF20500 TF20250
103258-222EA 325.91 USD InStock
103258-222 103258-220 103258-224
3D-Fect™ Transfection Reagent, OZ Biosciences
Transfection Reagents
3D-Fect is specifically developed to directly transfect cells cultured in 3D scaffolds.
  • Highly efficient: cell lines and primary cells
  • Specific for 3D Scaffolds
  • Compatible with all types of nucleic acids
  • Completely biodegradable
  • Long term protein expression
  • Serum Compatible
This 3D Transfection Reagent is suitable for all kinds of scaffolds and cells.
Delivery information: Shipped at room temperature
Packaging: Store at +4 °C
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty