Would you like to log in again?
3D-FectIN™ Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
3D-FectIN™
TN30500 TN30250 TN31000
103258-230EA 637.28 USD InStock
103258-230 103258-226 103258-228
3D-FectIN™ Transfection Reagent, OZ Biosciences
Transfection Reagents
3D-FectIN is the newest 3D-Transfection Reagent specifically developed to directly transfect cells cultured in 3D hydrogels.
  • Highly efficient: cell lines and primary cells
  • Specific for 3D Hydrogels
  • Compatible with all types of nucleic acids
  • Completely biodegradable
  • Long term protein expression
  • Serum compatible
3D-FectIN is suitable for all kind of hydrogels and cells.
Delivery information: Shipped at room temperature
Packaging: Store at +4 °C
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty