Would you like to log in again?
Viro-MICST™ Transduction Reagent, OZ Biosciences
Supplier: OZ Biosciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Viro-MICST™
VMX250 VMX1000 VMX500
103258-256EA 662.71 USD InStock
103258-256 103258-254 103258-406
Viro-MICST™ Transduction Reagent, OZ Biosciences
Transfection Reagents
Viro-MICST Transduction Reagent allows genetically modifying cells directly on magnetic cell purification columns.
  • Highly efficient
  • Rapid, simple, and ready to use
  • Suitable for all cells
Based on the i-MICST Technology (Integrated Magnetic Immuno-Cell Sorting and Transfection/Transduction), this technology combines cell isolation and genetic modification in one simple and reliable integrated system.
Delivery information: Shipped at room temperature
Packaging: Store at +4 °C
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty