Would you like to log in again?
Mag4C-LV Transfection Trial Kit, OZ Biosciences
Supplier: OZ Biosciences
Mag4C-LV
LTK11200 LKC11000
103258-150EA 205.38 USD InStock
103258-150 103258-258
Mag4C-LV Transfection Trial Kit, OZ Biosciences
Transfection Reagents
Mag4C-LV is designed for magnetic virus concentration
  • Simple, Rapid & Ready-to-use
  • Concentration of lentiviruses by magnetic capture in 30-45 minutes
  • High yield of viral capture and recovery
  • No need to process magnetic beads before capture
  • Avoid ultracentrifugation, precipitation and chemicals. Reduced handling steps of viruses
  • Suitable for all conditions & viruses
  • Improve virus stability under storage conditions
Mag4C-LV magnetic nanoparticles capture viruses in culture media with 80-99 % efficiency. Once captured onto magnetic beads, viruses can be:
Concentrated and stored with the Storage Buffer or directly used for downstream assay.
Concentrated, eluted from the magnetic beads with the Elution Buffer and stored with the Storage Buffer or used for subsequent assays.
This kit is specifically developed for capturing, concentrating and storing lentiviruses and retroviruses. This kit is composed of 3 reagents allowing Magnetic Capture/Concentration, Elution and Conservation of Lentiviruses and Retroviruses.
Delivery information: Shipped at room temperature
Packaging: Store at +4°C
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty