Would you like to log in again?
Pro-DeliverIN™ Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Pro-DeliverIN™
PI10100 PI11000 PI10250 PI10500
103258-304EA 390.12 USD InStock
103258-304 103258-302 103258-308 103258-306
Pro-DeliverIN™ Transfection Reagent, OZ Biosciences
Transfection Reagents
Pro-DeliverIN Transfection Reagent is an innovative reagent allowing intracellular delivery of biologically active proteins.
  • Intracellular delivery of functionally active proteins
  • Highly efficient in many primary cells and cell lines
  • Serum compatible
  • Biodegradable and non toxic
  • Straightforward protocol and ready-to-use reagent
This lipid-based formulation is the first serum compatible reagent to deliver functional proteins into living cells. The proteins delivered inside cells retain their biological functions.
Delivery information: Shipped at room temperature
Packaging: Store at +4 °C
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty