Would you like to log in again?
CPRG assay kit, OZ Biosciences
Supplier: OZ Biosciences
GC10002
103258-320EA 324.69 USD InStock
103258-320
CPRG assay kit, OZ Biosciences
Assays Enzyme Assays
OZ Biosciences CPRG assay kit is ideal to quantify low expression level of beta-Galactosidase
  • Convenient for all transfection assays
  • Suitable for cultured cells and tissues
  • Simple, rapid and easy-to-use
  • Economical
  • Optimized with stable buffers
The levels of active beta-Gal actosidase expression can be quickly measured by its catalytic hydrolysis of CPRG (Chlorophenol red-b-D-galactopyranoside), substrate to a dark red product. The high sensitivity of this substrate improves the measurement of beta-Galactosidase activity when the reporter gene expression is low. (Absorbance at 570-595 nm). All the necessary reagents are provided in this assay kit and offer a rapid, simple and sensitive method to quantify spectrophotometrically the enzyme expression level in LacZ transfected cells.
LacZ is one of the most frequently reporter gene used in transfection experiments. The LacZ encoded protein (beta-galactosidase, b-Gal), is very stable, resistant to proteolytic degradation and easily assayed. 
Delivery information: Shipped with a gel pack
Packaging: Store all reagents at +4°C except Beta Gal Enzyme and CPRG Substrate stock solution
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty