Would you like to log in again?
NeuroMag™ Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
NeuroMag™
NM51000 KC30800 NM50500 NM50200
103258-382EA 871.09 USD InStock
103258-382 103258-376 103258-378 103258-380
NeuroMag™ Transfection Reagent, OZ Biosciences
Transfection Reagents
NeuroMag Transfection Reagent is the first dedicated Magnetofection transfection reagent for neurons and neural cells, from 1 DIV to 21 DIV.
  • Great efficiency, ideal for primary neurons
  • Efficient from 1 DIV to 21 DIV
  • Non toxic and completely biodegradable: high transfected neurons viability
  • Ready-to-use, straightforward and rapid
  • For all types of nucleic acids
Accessories information: This reagent needs to be used with a magnetic plate.
Delivery information: Shipped at room temperature
Packaging: Store at -20 °C
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty