Would you like to log in again?
DreamFect™ Stem Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
DreamFect™
ST31000 ST30500
103258-394EA 477.35 USD InStock
103258-394 103258-392
DreamFect™ Stem Transfection Reagent, OZ Biosciences
Transfection Reagents
DreamFect Stem Transfection Reagent is a powerful reagent allowing multipotent stem cells transfection with high efficiency and very low toxicity.
  • High transfection efficiency for multipotent Stem cells
  • Minimized toxicity due to reagent biodegradability and low DNA amount required
  • Cell phenotype and differentiation potential are not affected
  • Serum Compatible
  • Simple, Ready-to-use and rapid (no specific buffer)
Delivery information: Shipped at room temperature
Packaging: Store at -20 °C
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty