Would you like to log in again?
EcoTransfect™ Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
EcoTransfect™
ET10500 ET13000 ET11000
103258-398EA 256.83 USD InStock
103258-398 103258-396 103258-400
EcoTransfect™ Transfection Reagent, OZ Biosciences
Transfection Reagents
EcoTransfect Transfection Reagent is an economical Lipofection reagent dedicated to the transfection of popular cell lines.
  • Economical
  • Good efficiency
  • Serum compatible
  • Simple, ready-to-use and rapid
It is the perfect solution to quickly analyze the biological activity of your nucleic acids, to perform routine transfection assays at low cost and to accomplish high throughput screening.
Delivery information: Shipped at room temperature
Packaging: Store at +4 °C
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty