Would you like to log in again?
pVectOZ™ Transfection Plasmids, OZ Biosciences
Supplier: OZ Biosciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
pVectOZ™
PL00030 PL00040 PL00150 PL00120 PL00050 PL00020 PL00010 PL00140 PL00130 PL00110
103258-174EA 405.86 USD InStock
103258-174 103258-172 103258-178 103258-420 103258-176 103258-330 103258-424 103258-422 103258-426 103258-180
pVectOZ™ Transfection Plasmids, OZ Biosciences
Transfection Reagents
pVectOZ Transfection Plasmids are engineered in an optimized backbone and are ideal for all transfections.
  • Highest levels of transgene expression in mammalian cells and tissues
  • Suitable for all transfection applications (in vivo and in vitro)
  • High copy number production in Escherichia coli
  • LPS-endotoxin free, supercoiled and highly purified: transfection grade approved
OZ Biosciences proposes five expressions vectors encoding for the most popular reporter genes (CAT, GFP, LacZ, Luciferase, SEAP).
Delivery information: Shipped at room temperature
Packaging: Store at -20 °C
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty