Would you like to log in again?
COSFect™ Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
COSFect™
CF10500 CF12500 CF11000
103258-436EA 404.66 USD InStock
103258-436 103258-390 103258-438
COSFect™ Transfection Reagent, OZ Biosciences
Transfection Reagents
COSFect is a lipid based transfection reagent specifically developed for COS cell transfection.
  • Highly efficient with COS cell lines
  • Ready-to-use: no need for additional buffer
  • Low nucleic acid amount - minimized toxicity
  • High level of nucleic acid compaction
  • Easy and straightforward protocol
  • Compatible with any culture medium: medium changed not required
The cationic design of COSFect reagent allows high nucleic acid compaction for a high transfection efficiency into COS cells.
Delivery information: Shipped at room temperature
Packaging: Store at -20 °C
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty