Would you like to log in again?
Verofect™ Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Verofect™
VF61000 VF60250 VF65000 VF60500
103258-112EA 345.29 USD InStock
103258-112 103258-402 103258-446 103258-444
Verofect™ Transfection Reagent, OZ Biosciences
Transfection Reagents
VeroFect Transfection Reagent is a powerful Lipofection reagent designed to obtain highly efficient and reproducible transfection of Vero cells.
  • Optimized for Vero cells
  • Non-toxic
  • Serum compatible
  • Simple, ready-to-use, and rapid
  • Highly efficient and reproducible results
It can be used for many applications such as stable and transient transfection, protein and viral production, etc.
Delivery information: Shipped at room temperature
Packaging: Store at +4 °C
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty