Would you like to log in again?
FlyFectIN™ Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
FlyFectIN™
FF50500 FF51000 FF55000
103258-450EA 402.23 USD InStock
103258-450 103258-452 103258-448
FlyFectIN™ Transfection Reagent, OZ Biosciences
Transfection Reagents
FlyFectin Transfection Reagent is a powerful reagent based on the TEE-technology and specifically designed to obtain highly efficient and reproducible transfection of insect cells.
  • Dedicated to insect cells
  • Ideal for production of recombinant protein using Baculovirus expression system.
  • Non toxic
  • Serum compatible
  • Simple, Ready-to-use and Rapid
It is adapted to the delivery of all types of nucleic acids.
Delivery information: Shipped at room temperature
Packaging: Store at +4 °C
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty