Would you like to log in again?
LipoMag™ Transfection Kit, OZ Biosciences
Supplier: OZ Biosciences
LipoMag™
LM80500 LM80250 LM80000
103258-454EA 1005.6 USD InStock
103258-454 103258-456 103258-218
LipoMag™ Transfection Kit, OZ Biosciences
Transfection Reagents
LipoMag Transfection Kit associates our most efficient lipid-based transfection reagent, DreamFect Gold with CombiMag magnetic nanoparticles, the most versatile Magnetofection reagent.
  • Highest efficiency without toxicity
  • Superior transgene expression level than any other reagents
  • Enhancement of Dreamfect™ Gold efficiency and outperformed competitors
  • Biodegradable
  • Multipurpose (various types of nucleic acids)
  • Universal (cell lines and primary cells)
  • Simple, ready-to-use and rapid
  • Serum compatible
By taking advantage of the Lipofection and Magnetofection transfection techniques, LipoMag Kit reaches even higher transfection efficiency especially in primary cells while reducing toxicity.
Accessories information: This kit needs to be used with a magnetic plate
Delivery information: Shipped at room temperature
Packaging: Store DreamFect Gold -20°C and CombiMag at +4°C
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty