Would you like to log in again?
NEBNext® Multiplex Oligos for Illumina (Index Primers Set 3), New England Biolabs
Supplier: New England Biolabs
NEBNext®
E7710S E7710L
103258-564EA 419 USD InStock
103258-564 103258-544
NEBNext® Multiplex Oligos for Illumina (Index Primers Set 3), New England Biolabs
Nucleic Acid Reagents Next Generation Sequencing Reagents
NEBNext Multiplex Oligos provide adaptors and primers to enable high yield multiplex Illumina library production. The unique hairpin loop structure of the NEBNext Adaptor minimizes adaptor-dimer formation, and NEBNext index PCR primers enable index incorporation during library amplification. Additional NEBNext Multiplex Oligos products are available for different indexing strategies.
  • High efficiency adaptor ligation
  • Minimized adaptor-dimer formation
  • Includes index primers for library multiplexing
  • Increased sample identification specificity
  • Includes USER Enzyme
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty