Would you like to log in again?
NEBNext® Ultra II DNA Library Prep with Sample Purification Beads, New England Biolabs
Supplier: New England Biolabs
NEBNext® Ultra™
E7103S E7103S E7103L E7103L
103258-560EA 678 USD InStock
CA103258-560 103258-560 CA103258-558 103258-558
NEBNext® Ultra II DNA Library Prep with Sample Purification Beads, New England Biolabs
Nucleic Acid Reagents Next Generation Sequencing Reagents
The NEBNext Ultra II DNA Library Prep with Sample Purification Beads contains the enzymes, buffers and purification beads required to convert a broad range of DNA input amounts into indexed libraries for next-generation sequencing on the Illumina platform.
  • Get more of what you need, with the highest library yields
  • Use to generate high quality libraries from a broad range of input amounts, from 500 pg to 1 µg
  • Prepare libraries from ALL your samples, including GC-rich DNA and FFPE DNA samples
  • Improve yields and quality for target enrichment applications
  • Includes SPRIselect beads
The fast, user-friendly workflow has minimal hands-on time and, for added convenience, this kit contains NEBNext Sample Purification Beads (SPRIselect beads from Beckman Coulter) for size selection and enzyme reaction cleanup.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty