Would you like to log in again?
Lysobacter Enzymogenes Endoproteinase LysC
Supplier: New England Biolabs
P8109S P8109S
103258-578EA 205 USD InStock
103258-578 CA103258-578
Lysobacter Enzymogenes Endoproteinase LysC
Enzymes
Endoproteinase LysC is a serine endoproteinase, isolated from Lysobacter enzymogenes, that cleaves peptide bonds at the carboxyl side of lysine. LysC is a sequencing grade enzyme and is suitable for proteomics and glycobiology applications.
  • Analyze complex proteomes
  • Compatible for simultaneous co-digestion with Trypsin-ultra Mass Spectrometry Grade
  • Suitable for both in-gel and solution digests
  • Supplied lyophilized
  • Optimal activity and stability for up to 12 months
Delivery information: Supplied freeze dried from Tris-HCl buffer, pH 8.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty