Would you like to log in again?
Cell Staining Buffer, Southern Biotechnology
Supplier: Southern Biotechnology
0225-01L 0225-01M 0225-01S
103259-764EA 108.54 USD InStock
103259-764 103259-766 103265-564
Cell Staining Buffer, Southern Biotechnology
Buffers Microscopy Buffers
Cell Staining Buffer maximizes fluorescent signal intensity generated by pH sensitive fluorochromes.
  • Maximizes fluorescence signal intensities
  • Minimizes non-specific binding
  • Useful for the dilution fluorescent reagents as well as for the suspension, washing, and storage of cells for flow cytometry
Designed to maximize fluorescence signal intensities generated by pH-sensitive fluorochromes. In addition, this buffer contains a metabolic inhibitor sodium azide (NaN3) and animal serum proteins to help minimize non-specific binding. NaN3 helps inhibit patching and capping of cell surface antigens. It also inhibits the potential redistribution of cell surface antigens caused by antibody cross linking, thereby preventing the potential loss of fluorescent signal intensities generated by immunofluorescently-stained cells during subsequent flow cytometric analysis.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty