Would you like to log in again?
SEM Mount, For Hitachi, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
75346-H 75344-H 75355-H
103301-900PK 15.41 USD InStock
103301-900 103301-902 103301-904
SEM Mount, For Hitachi, Electron Microscopy Sciences
Specimen Mounting Supplies
Hitachi SEM mounts are complete line of SEM specimen mounts are made from lathe finished ultra-pure aluminum.
  • For scanning electron microscopy
  • Standard lathe finish are economically priced
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty