Would you like to log in again?
Forensic Gunshot Residue Field Kits, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
76480-11 76480-15 76480-13
103301-914KT 29 USD InStock
103301-914 103302-464 103302-484
Forensic Gunshot Residue Field Kits, Electron Microscopy Sciences
Specimen Mounting Supplies
Gun Shot Residue Kit for Scanning Electron Microscopy.
  • Powder-free nitrile gloves
  • Evidence label
  • Chain of custody label on box
  • Tamper-evident cardboard transport box (41/2 W x 33/4 D x 3" H)
  • Tamper evident seal
The certified SEM pin stubs have very low amounts of Pb, Sb and Ba, not detectable by SEM/EDX. The caps securely hold the GSR sample disc for easy GSR collection. Pre-labeled discs and tubes are both ready-for-sampling. 3-Disc GSR Field Kit has two sampling discs plus a control disc to enhance certainty of the analysis. 4-Disc GSR Field Kit is useful for sampling palm and back separately on both hands. 5-Disc GSR Field Kit is compatible with FBI requirements, with four discs plus a control disc.
Delivery information: Kit includes certified SEM pin stubs with double-coated adhesive carbon tabs in pre-labelled plastic tubes. A sturdy, sealed tamper evident transport box is also included.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty