Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Developer D-19 Powder Replacement, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
74205 74210 74200
100488-596PK 52.64 USD InStock
100488-596 100499-934 103301-918
Developer D-19 Powder Replacement, Electron Microscopy Sciences
Film Processing Supplies
A high contrast developer for electron microscopy films.
Delivery information: Kit contains pre-weighed amounts of: Distilled Water (48 °C/125 °F), Metol, Sodium Sulfate, Hydroquinone, Sodium Carbonate (monohydrate), Potassium Bromide to make 1 liter or 1 gallon of developer.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty