Would you like to log in again?
Forensic Gunshot Residue Lab Kit, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
76480-22 76480-19 76480-18
103301-944KT 29.87 USD InStock
103301-944 103302-390 103301-976
Forensic Gunshot Residue Lab Kit, Electron Microscopy Sciences
Specimen Mounting Supplies
SEM mounts with adhesive tab secures in glass vial to avoid contamination.
  • Certified aluminum SEM mount
  • Set of 10 in transport box
  • Vials are 25 OD × 55 i mm high
This Kit is designed to avoid any contamination of GSR samples. Also available as a sample kit without mounts and tabs.
Delivery information: Includes ten 12.7 mm SEM pin mounts in clean, glass storage tubes (25 mm OD x 55 mm high) secured in a compact transport box with labels.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty