Would you like to log in again?
Dumont Tweezer Handles, Style WA, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
10004-WA2-SM 10004-WA1-SM 10004-WA3-SM
103301-968EA 53.46 USD InStock
103301-968 103301-970 103301-972
Dumont Tweezer Handles, Style WA, Electron Microscopy Sciences
Forceps
These precision tweezers from Dumont made of high quality steels with perfect finish suits all application needs.
These tweezer handles are anti-magnetic. Can be used with all tips with four screws. Antimagnetic (80%) contains C, Cr, Ni and Mo and has good resistance to corrosion due to a high concentration of molybdenum but is not as hard as Inox.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty