Would you like to log in again?
ROR (Residual Oil Remover), Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
74319-30 74319-35
103302-468EA 15.61 USD InStock
103302-468 103302-020
ROR (Residual Oil Remover), Electron Microscopy Sciences
Cleaning Agents Detergents and Surface Cleansers
Residual Oil Remover is designed to emulsify and remove microscopic oil residues a by-product of environmental pollution from the surface and pores of glass and plastic.
  • Increases brightness and clarity
  • Safe and versatile for delicate surfaces
  • Available in either a dropper bottle or a Pump style bottle
ROR has been tested and used on everything from multicoated lenses to beryllium surfaced contacts and has proven time and again to be the finest and safest optical cleaning treatment.
ROR is used today by the military, NASA, astronomers, scientists, hospitals, laboratories and universities.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty