Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Membrane Boxes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64304-23 64304-58 64304-42 64304-39 64304-56 64304-13 64304-16 64304-46 64304-45 64304-17 64304-19 64304-44 64304-38 64304-25 64304-30 64304-20 64304-57 64304-43 64304-40 64304-28 64304-55 64304-21 64304-51 64304-27 64304-24 64304-18 64304-14 64304-29 64304-36 64304-12 64304-22 64304-59 64304-50 64304-26 64304-15 64304-11 64304-10 64304-41 64304-52 64304-54
100498-658EA 40.12 USD InStock
100498-658 101412-938 100498-656 103301-958 101412-934 101412-956 101412-958 101412-936 103302-336 103302-654 103302-038 103302-338 101412-930 101412-952 103302-350 101412-954 101412-932 103302-036 100490-554 103302-034 103302-352 101412-946 103301-962 101412-948 100498-662 100498-660 103301-960 103302-402 103302-620 103302-528 103302-406 103302-404 101412-942 103302-382 101412-944 101412-960 103302-420 103302-386 101412-940 101412-962
Membrane Boxes, Electron Microscopy Sciences
Mailers and Shippers
These unique membrane boxes are for the transfer, storage and shipping of many delicate items. Once your sensitive item is placed in the box the elastic membrane conforms around the enclosed object and protects it from any movement.
  • Protects items from any movement
  • Double elastic membrane on the top and bottom offers security
  • Available in round, rectangular and square shapes
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty