Would you like to log in again?
Tweezers WA2 Handle with CA275-CW Tips, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
TIP-CA275-CW 10004-CA275-SM
103301-864EA 12.13 USD InStock
103301-864 103302-054
Tweezers WA2 Handle with CA275-CW Tips, Electron Microscopy Sciences
Forceps
These tweezers have an anti-magnetic WA2 handle with replaceable CA275-CW tips.
  • Hand made of the highest quality steels and alloys
  • Tips attach with screws
  • High precision and perfectly finished to suit the application
Antimagnetic (80%) contains C, Cr, Ni and Mo and has good resistance to corrosion due to a high concentration of molybdenum but is not as hard as inox.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty