Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Biopsy Bags, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

62326-10 62326-05 62326-03 62326-01 62326-02 62326-15
103302-090PK 89.94 USD InStock
103302-090 100490-170 100501-992 103302-474 103302-550 103301-906
Biopsy Bags, Electron Microscopy Sciences
Bags Sample Bags
Bags are made up of porous paper which provides maximum tissue safety during processing.

  • Made from porous paper
  • Permits proper fluid exchange and drainage
  • Available in 25 and 35 mm diameter or 25 and 32 mm sizes

They permit proper fluid exchange and drainage.

To use: simply introduce the specimen into the bag and fold it enclosing the cassette or capsule. Now ready to process as usual.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty