Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Clearium® Mounting Media, Electron Microscopy Science
Supplier: Electron Microscopy Sciences
Clearium®
13520-13 13520-12 13520-11
103306-390CS 371.57 USD InStock
103306-390 103302-098 103306-056
Clearium® Mounting Media, Electron Microscopy Science
Stains Microscopy and staining reagents
Clear and coverslip your slide preparations in one easy step.
  • Eliminates need for hazardous xylene or xylene substitutes
  • Eliminates final step of clearing
  • Coverslip directly from absolute isopropyl alcohol
This unique product eliminates the need for hazardous xylene or xylene substitutes as a clearing agent in histology and cytology laboratories. With Clearium®, simply do away with the final step of clearing and coverslip directly from absolute isopropyl alcohol.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty