Would you like to log in again?
Athene Square Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
A200-NI A300-CU A150-CU A400-CU A300-NI A50-CU A200-CU
103302-456PK 56.31 USD InStock
103302-456 103302-094 103302-646 103302-118 103302-120 103302-096 103302-122
Athene Square Grids, Electron Microscopy Sciences
Graticules Reticles and Grids
Highest quality square mesh transmission electron microscope grids.
  • Copper and nickel grids
  • 50, 150, 200, 300, and 400 mesh grids
  • Good handling characteristics
Individually inspected before being packed.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty