Would you like to log in again?
SEM Clip Specimen Mounts, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
75923-20 75923-22 75923-14 75923-15 75923-21 75923-12 75923-10 75923-23 75923-16 75923-19 75923-13 75923-11 75923-18 75923-17
103301-950EA 41.07 USD InStock
103301-950 103302-546 103302-458 103302-368 103302-114 103302-384 103302-460 103302-370 103302-124 103302-112 103301-948 103302-342 103302-044 103302-462
SEM Clip Specimen Mounts, Electron Microscopy Sciences
Specimen Mounting Supplies
Specially designed for easy, quick and clean mounting of any type of thin specimen on specimen stubs.
  • Available with single and multiple clips
  • Ideal for holding silicon chips, paper, wire, threads, thin films, sheet metal, and more
  • SEM clip mounts with multiple clips can hold larger samples or multiple smaller samples
These spring-loaded clips accommodate samples of up to 2 mm thickness, samples are held securely and allow for easy change of samples. Adhesives are not required, eliminating possible outgassing issues and saving time.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty