Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Cylinder Mount Hitachi M4, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
75954-25 75930-05 75954-15 75930-35 75930-15 75954-35 75930-10 75954-30 75930-25 75930-20 75954-40
103302-128EA 74.33 USD InStock
103302-128 103302-622 103302-534 103302-688 103302-126 103302-506 103304-382 103302-504 103302-372 103302-694 103302-374
Cylinder Mount Hitachi M4, Electron Microscopy Sciences
Specimen Mounting Supplies
Spring-loaded clips hold specimens for scanning electron microscopy without adhesives.
  • Spring loaded clips accommodate samples up to 2 mm thickness
  • Available with single and multiple clips
  • Available in 25, 32, or 50 mm diameter
Samples are held securely by small spring-loaded clips and allow for easy change of samples. Adhesives are not required, eliminating possible outgassing issues and saving time. Ideal for holding silicon chips, paper, wire, threads, thin films, sheet metal and more. SEM Clip Mounts with multiple clips can hold larger samples or multiple smaller samples.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty