Would you like to log in again?
EMS Shield Mounting Medium with DAPI and DABCO, Electron Microscopy Science
Supplier: Electron Microscopy Sciences
17989-21 17989-20
103302-252EA 155.17 USD InStock
103302-252 103306-188
EMS Shield Mounting Medium with DAPI and DABCO, Electron Microscopy Science
Stains Microscopy and staining reagents
EMS Shield with DAPI is an aqueous mounting medium for preserving fluorescence of tissue and cell smears.
  • Does not contain phenylenediamine, which destroys immunofluorescence of Cy dyes, RP-E, PC and APC
  • Fortified with DAPI a counter-stain for DNA
  • Used in situ hybridization techniques or other methods where fluorescence of DNA staining is required
  • RNA is also stained with DAPI
EMS Shield with DAPI is an aqueous mounting medium for preserving fluorescence of tissue and cell smears. This unique formula prevents rapid photobleaching of FITC, Texas Red, AMCA, Cy2, Cy3, Cy5, Alexa fluoro 488, Alexa fluoro 594, Green fluorescent protein (GFP), tetramethyly rhodamine, Redox. Phycoerythrin (RP-E), Phycocyanin (PC), and Allophycocyanin (APC). Fluorescence is retained during prolonged storage at 4°C in the dark. This medium does not contain phenylenediamine, which destroys immunofluorescence of Cy dyes, RP-E, PC and APC.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty