Would you like to log in again?
Microbead Bag, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68605-20
103302-322EA 24.18 USD InStock
103302-322
Microbead Bag, Electron Microscopy Sciences
Bags Storage Bags
For using and storing thermal microbeads.
  • Designed to fit ice buckets
  • Use on top of cool bricks for prolonged cooling
Nylon mesh bag with drawstring closure for storing, cleaning, using, and transporting thermal microbeads. The bag is designed to store microbead in the freezer until ready for use. The bags fits into the 5 L ice bucket.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty