Would you like to log in again?
Earth-Friendly Long Handle Sampling Spoons, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72132 72131 72133
103301-936PK 33.86 USD InStock
103301-936 103302-340 103302-330
Earth-Friendly Long Handle Sampling Spoons, Electron Microscopy Sciences
Spoons
Long handles ideal for acquiring samples from bottles, jars, and more.
  • Length 17.8 cm
  • Non-sterile
  • Translucent
  • Available in 3 bowl sizes
Long-handled spoons are ideal for acquiring samples from bottles, jars, and other confined areas and helping to avoid personal contact with sample materials. A flat edge on the rim of each bowl allows fast levelling to collect and measure small amounts with accuracy. Each spoon is 17.8 cm (7") overall length. Non-sterile and translucent in color.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty