Would you like to log in again?
Double Slot Mini Vise, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
75951-03
103302-378EA 42.81 USD InStock
103302-378
Double Slot Mini Vise, Electron Microscopy Sciences
Specimen Mounting Supplies
Clamp thin specimens and cross sections without conductive paint or tape for viewing in a scanning electron microscope using the double slot vise specimen holder.
  • Use for paper, metals, plastic, textiles, plant material and more
  • Machined aluminum with stainless steel allen set screws
Pin length: 3.2 mm (1/8")
1 mm wide x 3 mm deep slots
Delivery information: Includes allen wrench.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty