Would you like to log in again?
Plexiglas Microscope Slides, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71891-10
103302-388EA 58.36 USD InStock
103302-388
Plexiglas Microscope Slides, Electron Microscopy Sciences
Slides Microscope Slides
A Fool-Proof method for the mounting of serial sections on Single Hole Grids.
They prevent wrinkling and section loss while mounting sections on the grid.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty