Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Stainless Steel Baskets with Multiple Compartments, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70194-29 70194-26 70194-28 70194-27
103302-670EA 79.32 USD InStock
103302-670 103302-344 103302-398 103302-066
Stainless Steel Baskets with Multiple Compartments, Electron Microscopy Sciences
Baskets Safety Transporting Baskets
These stainless steel baskets with lids and multiple compartments are used for critical point drying.
  • Single or multiple compartments
These baskets are either 15 or 23 mm in diameter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty