Would you like to log in again?
Tweezers Essence, WA1 Handle with 45-BUIS Tips, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
10004-B45-SM TIP-B45-BUI
103302-416EA 55.13 USD InStock
103302-416 103303-320
Tweezers Essence, WA1 Handle with 45-BUIS Tips, Electron Microscopy Sciences
Forceps
These precision tweezers from Dumont made of high quality steels with perfect finish suits all application needs.
These tweezers have an anti-magnetic WA1 handle with replaceble 45-BUIS tips made of boxwood (Buxus). Tips are attached with screws. Torque: cN.m 9 ±1. Length is 130mm.
Antimagnetic (80%) contains C, Cr, Ni and Mo and has good resistance to corrosion due to a high concentration of molybdenum but is not as hard as Inox.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty