Would you like to log in again?
Rotary Diamond Micro-Engraver Pen, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
50465-15
103302-470EA 37.66 USD InStock
103302-470
Rotary Diamond Micro-Engraver Pen, Electron Microscopy Sciences
Pens Stylus and Scribers
This micro-engraver pen may be used to mark samples, engrave tools, indicate a sample area for finding in the SEM and deburring.
  • Easy to handle and control, this micro engraver is ideal for detailed engraving on almost any surface like wood, metal, ceramic and glass
  • Contoured grip for comfort and control
  • Pen-type design fits in a shirt pocket or purse and goes anywhere
  • Precision-ground stainless steel bit with diamond point tip
  • Requires two AAA batteries
The pen measures 61/4" and weighs 0.8 kg.
Delivery information: Two AAA batteries are included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty