Would you like to log in again?
Eyelash Manipulator; An EM Tool, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71182
103302-486PK 22.43 USD InStock
103302-486
Eyelash Manipulator; An EM Tool, Electron Microscopy Sciences
Microtomes and Microtome Assemblies Blades and Handles
This is for direct manipulation of sections and small tissue samples.
  • Sable brush handle made from the highest quality wood
  • Cleaned and sterilized with alcohol and a tissue
Suitable for ultramicrotomy to carefully tease sections that are in the diamond knife boat.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty