Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SEM Large Double Slot Vise, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

75951-04
103302-508EA 55.27 USD InStock
103302-508
SEM Large Double Slot Vise, Electron Microscopy Sciences
Specimen Mounting Supplies
Clamp thin specimens and cross sections without conductive paint or tape for viewing in a scanning electron microscope using the double slot vise specimen holder.

  • Use for paper, metals, plastic, textiles, plant material and more
  • Machined aluminum with stainless steel allen set screws

2.5 mm wide x 5 mm deep slots
Dimensions: 25 x 8 mm (1 x 0.32") with 3.2 mm (1/8") pin

Delivery information: Includes allen wrench.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty