Would you like to log in again?
Gold Coated Tweezers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
78750-01 78750-12 78750-07 78750-09 78750-11 78750-04 78750-06 78750-10 78750-05 78750-08 78750-03 78750-02
103302-138EA 67.65 USD InStock
103302-138 103302-644 103302-446 103302-610 103302-676 103302-532 103302-136 103301-810 103302-448 103302-542 103302-144 103301-818
Gold Coated Tweezers, Electron Microscopy Sciences
Forceps
These gold coated tweezers are finely crafted, high precision tweezers made from anti-acidic, non-magnetic stainless steel, plated with 2 microns of pure 24-carat gold
  • Resists chemical corrosion and oxidation
  • Excellent conductor of electricity
These tweezers are autoclavable and conducive for use when working with microelectronics, TEM and immunogold staining, electro-chemistry and nanotechnology.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty