Would you like to log in again?
Thin Specimen Split Mount on Pin Stub, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
75950-08
103302-548EA 60.14 USD InStock
103302-548
Thin Specimen Split Mount on Pin Stub, Electron Microscopy Sciences
Specimen Mounting Supplies
Thin specimen split mount on pin stub is used for cross section examination in scanning electron microscopes.
  • Used for paper, metals, plastic, textiles, plant material and more
  • Machined aluminum with stainless steel allen set screws
  • Opens to 6.4 mm
  • Grooved head
Height of head is 7.4 mm (0.29"), Pin Length is 8 mm (0.314") and pin is centered.
Delivery information: Includes allen wrench.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty