Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Thin Specimen Split Mount (AMRAY), Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences
75950-09
103302-626EA 68.86 USD InStock
103302-626
Thin Specimen Split Mount (AMRAY), Electron Microscopy Sciences
Specimen Mounting Supplies
For use in Amray electron microscopes

15 mm (0.59") dia., 3.2 mm (⅛") dia. pin.
Opens to 3.75 mm (⅜").
Pin length: 14.3 mm (9/16").
Height of head: 10.2 mm (0.4"), centered split. Pin is off-center.
Material: Machined aluminum with two stainless steel allen set screws.

Delivery information: Includes allen wrench.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty