Would you like to log in again?
Earth-Friendly Sticks, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72143-10 72144-11
103302-652EA 85.64 USD InStock
103302-652 103306-374
Earth-Friendly Sticks, Electron Microscopy Sciences
Spatulas
Earth friendly sticks are used for mixing samples, handling capsules, scraping containers, and positioning solid samples under examination.
The sticks measure 14.6 x 1.9 x 0.15 cm (53/4 x 3/4 x 1/16"). Translucent.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty