Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Muscovite Mica Sheets, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71857-15 71856-01 71858-01 71856-01-10 71855-10 71857-11 71856-02 71855-01 71857-10-10 71852-01 71857-11-10 71857-10 71855-01-10 71855-15-10 71853-15 71855-05 71855-10-10 71851-05 71850-15 71857-15-10 71850-11 71853-11 71850-10 71857-05-10 71855-15 71853-05 71854-15 71850-01 71853-10 71858-01-10 71856-04 71856-03 71857-01-10 71857-05 71854-01 71855-11-10 71857-01 71853-01 71855-05-10 71855-11
100493-514EA 3.09 USD InStock
100493-514 103301-956 100489-374 100489-372 100493-516 100493-518 100489-370 103301-954 100489-378 100493-510 100493-512 100489-376 100488-344 103302-314 100488-346 103302-312 103302-318 100499-576 103302-316 103302-472 103302-494 103302-692 103302-492 103302-696 103302-552 100489-384 100502-194 100489-382 100493-506 100493-508 100489-380 100502-192 100501-900 100491-016 100493-520 100489-368 100493-522 103302-346 103302-888 103302-320
Muscovite Mica Sheets, Electron Microscopy Sciences
Specimen Mounting Supplies
EMS Mica Sheets offer a clean surface for EM applications, carbon filming and particle spraying, as well as for AFM applications. The highest quality mica (V-1 or V-2) for AFM applications to the Medium Quality (V-4 to V-6) for replication and thin film deposition.
  • High Quality Muscovite Mica
  • EM applications, carbon filming and particle spraying and AFM applications
  • Peels off very thin up to 0 0001" uniform layers
There are two types of mica: muscovite and phlogopite. Generally, one differs from the other by color (Muscovite is Ruby, Green or White; Phlogopite is Amber, Yellow, or Silver). The maximum operating temperature for Muscovite is about 500 to 600 °C and for Phlogopite is about 800 to 900 °C. Our line consists of High Quality Muscovite Mica. This mica peels off very thin up to 0.0001" uniform layers, exposing "virgin" mica upon splitting.
Characteristics: Muscovite, potash type mica, sometimes known as granitic mica, is the best of all micas in dielectric strength, perfection of cleavage, and transparency. It has the following chemical formula: H2KAI3(SiO4)3. It has a vitreous luster and is colorless to gray, brown, pale green, violet, dark olive green, or rose red. It may be transparent to translucent. It has strong double refraction and is optically negative. It loses water of constitution at 600 °C and is practically non-magnetic. It exhibits pleochrism, which is the property of varying in color when viewed from different angles. RUBY muscovite is harder than green and has a pale brownish red color in thin sheets (0.020") or ruby red in thick plates (0.4"). Ruby mica can be split easily into films of 0.001" or thinner because it has such excellent cleavage. In other colors, this thinness can be obtained but at a considerable risk of cracking.
Quality Determination:
The quality of muscovite mica is verbal determination by visual quality classification ASTM (D351-57T) from the best V-1 to the worst is V-10A.
V-1: Clear - Hard, of uniform color, nearly flat, free of all stains, foreign inclusion, cracks, and other similar defects.
V-2: Clear and Slightly Stained - Hard, of uniform color, nearly flat and may contain slight crystallographic discoloration, and very slight air inclusions and not more than one fourth of the usable area.
V-4: Good Stained - Hard, of uniform color, may contain medium waves slight crystallographic discoloration, and medium air inclusion in not more than two-third of the usable area.
V-5: Stained A Quality - Hard, may contain medium air inclusions, uniformly distributed in the usable area; slight green vegetable stains, medium waviness, and heavy waves if specified.
Applications: In general, for those whom are using mica for the making of carbon support films, thin films coatings research, and some AFM studies where HOPG are used to taking place, the mica V-4 or V-5 is recommended. (Mica must always use freshly cleaved surfaces). For AFM and SPM calibration, V-1 or V-2 is recommended. However, we believe that it is important that you know what kind of mica is being used in your lab. The information on the mica as given above should be used to make your choice.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty