Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Lacey Carbon Film, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
LC400-CU-25 LC200-AU LC300-AU-150 LC300-NI-100 LC400-AU-100 LC200-AU-25 LC305-NI LC400-AU-25 LC200-AU-UL LC300-AU-25 LC400-NI-100 LC300-CU-UL LC325-CU-25 LC200-CU LC400-NI LC200-CU-100 LC200-CU-25 LC305-CU LC300-CU-150 LC200-AU-150 LC200-NI-100 LC400-CU-100 LC400-AU LC400-AU-UL LC305-AU LC400-NI-25 LC200-NI-UL LC400-CU-UL LC200-NI LC300-NI-150 LC200-NI-150 LC400-CU-150 LC400-AU-150 LC400-CU LC300-AU-100 LC400-NI-150 LC300-NI-UL LC300-AU LC325-AU-25 LC325-NI-25 LC300-AU-UL LC200-AU-100 LC200-NI-25 LC300-CU-100 LC200-CU-UL LC200-CU-150
103305-116PK 229.87 USD InStock
103305-116 76439-880 103305-118 103302-716 103305-112 103305-114 76439-834 103302-898 76439-878 100491-194 76439-932 103302-754 103302-896 102101-428 100491-192 103302-752 102101-448 102101-426 103302-714 102101-424 103302-712 102101-446 103302-890 102101-444 76439-918 102101-442 103302-894 103302-750 102101-440 103302-892 103305-120 76439-870 76499-776 100502-792 103305-122 100491-184 102101-438 100491-188 76439-862 103302-900 102101-436 102101-434 102101-452 102101-430 76439-848 102101-450
Lacey Carbon Film, Electron Microscopy Sciences
Graticules Reticles and Grids
Lacy carbon film provides support without interfering with transmission electron microscopy observation.
  • Copper, nickel and gold grids in 200, 300, and 400 mesh grids
  • Optically checked followed by batch testing in the EM
This carbon coated film on a broken pattern consists of woven-mesh-like holes of different sizes and shapes.
Ordering information: Molybdenum grids available on request.
Packaging: Packed in grid storage box.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty