Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
EDTA Buffer (10X), pH 8.5, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64142-10 64142-09
103305-700EA 267.81 USD InStock
103305-700 103302-792
EDTA Buffer (10X), pH 8.5, Electron Microscopy Sciences
Buffers Cloning Buffers
This buffer is intended for heat-induced antigen retriever on formalin-fixed paraffin-embedded (FFPE) tissue sections prior to application of antibodies.
In IHC most commonly used fixative like formalin mask tissue antigens (cellular, membrane and nuclear) by cross-linking process, this results in poor or no staining in IHC
Preparation of Working Solution: Dilute this 10X buffer as needed (90 mL of deionized or distilled water+10 mL of this buffer) mix well, 1X buffer can be stored at 2 to 8 °C.
The use of this buffer on FFPE tissue section improves accessibility of antibodies to tissue antigens.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty