Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Formvar Film on Hexagonal Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
FF300H-CU-UL FF400H-CU-TH FF400H-CU-ET FF100H-AU-TH FF400H-CU-UL FF200H-NI-25 FF100H-AU-ET FF200H-CU-50 FF300H-CU-50 FF200H-AU-TH FF300H-AU-25 FF100H-CU-25 FF100H-NI-TH FF300H-NI-50 FF400H-AU-50 FF200H-AU-UL FF300H-CU-TH FF300H-CU-25 FF200H-NI-TH FF100H-AU-50 FF400H-AU-TH FF200H-NI-50 FF200H-AU-25 FF200H-NI-ET FF200H-NI-UL FF300H-AU-ET FF300H-CU-ET FF400H-NI-ET FF200H-CU-TH FF200H-CU-UL FF300H-AU-UL FF100H-NI-UL FF200H-AU-50 FF200H-CU-25 FF100H-NI-50 FF400H-NI-TH FF300H-AU-TH FF100H-AU-25 FF400H-NI-25 FF100H-AU-UL FF100H-CU-ET FF400H-AU-UL FF100H-NI-25 FF300H-NI-TH FF200H-CU-ET FF300H-NI-ET FF300H-NI-25 FF400H-CU-25 FF200H-AU-ET FF100H-CU-TH FF400H-AU-ET FF100H-NI-ET FF400H-AU-25 FF300H-NI-UL FF300H-AU-50 FF100H-CU-UL FF400H-CU-50 FF400H-NI-50 FF400H-NI-UL FF100H-CU-50
103303-400PK 160.06 USD InStock
103303-400 102100-808 103303-402 103303-206 103303-404 103303-208 103302-238 102100-840 102100-762 102100-842 102100-764 102100-844 102100-800 102100-766 102100-802 102100-846 102100-768 102100-804 102100-848 102100-806 103303-510 103303-234 103303-512 103303-236 103303-230 103303-398 103303-232 102100-770 102100-772 103302-208 102100-850 103302-206 102100-852 102100-854 103302-240 102100-856 103302-244 103302-242 103302-182 103303-340 103303-100 103303-506 103303-508 103302-212 103302-210 103303-334 103303-212 103303-336 103303-338 103303-094 103303-096 103303-098 103303-210 102100-798 103302-186 103302-184 102100-834 102100-836 103302-188 102100-838
Formvar Film on Hexagonal Grids, Electron Microscopy Sciences
Graticules Reticles and Grids
A thin film of pure formvar resin coats the shiny side of a Hexagonal grid.
  • Formvar coatings thickness of standard (10 nm), ultra-thin (5 to 6 nm), thick (15 to 20 nm), or extra thick (25 to 50 nm)
  • Select from copper, nickel, and gold grids in 100, 200, 300, and 400 mesh sizes
Optically checked followed by batch testing in the EM.
Packaging: Packed in grid storage box.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty